Assay Details
| Gene Symbol: LOC4334276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775118.1 | XP_015630604.1 | 1 - 2 | 161 | 111 | |
| GenBank mRNA | AK065851.1 | - | 1 - 2 | 99 | 111 | |
| AK119532.1 | - | 1 - 2 | 151 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775118.1 transcripts.
Set Membership: |
|

LOC4334276
Genomic Map