Assay Details
| Gene Symbol: LOC4334312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775600.2 | XP_015631086.1 | 7 - 8 | 895 | 122 | |
| GenBank mRNA | AK100619.1 | - | 7 - 8 | 873 | 122 | |
More Information
Set Membership: |
|

LOC4334312
Genomic Map