Assay Details
Gene Symbol: LOC4334752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774202.1 | XP_015629688.1 | 2 - 3 | 2364 | 83 | |
XM_015774203.2 | XP_015629689.1 | 2 - 3 | 2364 | 83 | ||
XM_015774204.2 | XP_015629690.1 | 2 - 3 | 2364 | 83 | ||
GenBank mRNA | AK070364.1 | - | 2 - 3 | 2336 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
