Assay Details
| Gene Symbol: LOC4334900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778943.2 | XP_015634429.1 | 1 | 879 | 93 | |
| GenBank mRNA | AK110629.1 | - | 1 | 857 | 93 | |
More Information
Set Membership: |
|

LOC4334900
Genomic Map