Assay Details
| Gene Symbol: LOC4334928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777983.2 | XP_015633469.1 | 6 - 7 | 2446 | 130 | |
| XM_015777985.2 | XP_015633471.1 | 7 - 8 | 2356 | 130 | ||
| GenBank mRNA | AK063994.1 | - | 6 - 7 | 2437 | 130 | |
| AK073160.1 | - | 6 - 7 | 2352 | 130 | ||
More Information
Set Membership: |
|

LOC4334928
Genomic Map