Assay Details
| Gene Symbol: LOC4335102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778345.2 | XP_015633831.1 | 3 - 4 | 632 | 68 | |
| GenBank mRNA | AK063110.1 | - | 3 - 4 | 624 | 68 | |
More Information
Set Membership: |
|

LOC4335102
Genomic Map