Assay Details
| Gene Symbol: LOC4335541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778702.2 | XP_015634188.1 | 5 - 6 | 819 | 92 | |
| GenBank mRNA | AK061248.1 | - | 5 - 6 | 819 | 92 | |
| AK103339.1 | - | 5 - 6 | 775 | 92 | ||
More Information
Set Membership: |
|

LOC4335541
Genomic Map