Assay Details
| Gene Symbol: LOC4335655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780850.2 | XP_015636336.1 | 2 - 3 | 517 | 131 | |
| GenBank mRNA | AK067193.1 | - | 2 - 3 | 513 | 131 | |
| AK104891.1 | - | 2 - 3 | 471 | 131 | ||
More Information
Set Membership: |
|

LOC4335655
Genomic Map