Assay Details
| Gene Symbol: LOC4335655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780850.2 | XP_015636336.1 | 3 - 4 | 811 | 66 | |
| GenBank mRNA | AK067193.1 | - | 3 - 4 | 807 | 66 | |
| AK104891.1 | - | 3 - 4 | 765 | 66 | ||
More Information
Set Membership: |
|

LOC4335655
Genomic Map