Assay Details
| Gene Symbol: LOC4335655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780850.2 | XP_015636336.1 | 4 - 5 | 930 | 112 | |
| GenBank mRNA | AK067193.1 | - | 4 - 5 | 926 | 112 | |
| AK104891.1 | - | 4 - 5 | 884 | 112 | ||
More Information
Set Membership: |
|

LOC4335655
Genomic Map