Assay Details
| Gene Symbol: LOC4335786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781396.2 | XP_015636882.1 | 10 - 11 | 1565 | 78 | |
| GenBank mRNA | AK073418.1 | - | 10 - 11 | 1556 | 78 | |
More Information
Set Membership: |
|

LOC4335786
Genomic Map