Assay Details
| Gene Symbol: LOC4335786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781396.2 | XP_015636882.1 | 6 - 7 | 1115 | 75 | |
| GenBank mRNA | AK073418.1 | - | 6 - 7 | 1106 | 75 | |
More Information
Set Membership: |
|

LOC4335786
Genomic Map