Assay Details
| Gene Symbol: LOC9267691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780917.2 | XP_015636403.1 | 16 - 17 | 2588 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9267691| Gene Symbol: LOC9267691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780917.2 | XP_015636403.1 | 16 - 17 | 2588 | 71 | |
Set Membership: |
|