Assay Details
| Gene Symbol: LOC4336037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781427.2 | XP_015636913.1 | 4 - 5 | 602 | 94 | |
| GenBank mRNA | AK059553.1 | - | 4 - 5 | 528 | 94 | |
| AK104444.1 | - | 4 - 5 | 504 | 94 | ||
More Information
Set Membership: |
|

LOC4336037
Genomic Map