Assay Details
| Gene Symbol: LOC4336045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780503.2 | XP_015635989.1 | 6 - 7 | 762 | 82 | |
| XM_015780504.2 | XP_015635990.1 | 5 - 6 | 726 | 82 | ||
| GenBank mRNA | AK061516.1 | - | 6 - 7 | 668 | 82 | |
More Information
Set Membership: |
|

LOC4336045
Genomic Map