Assay Details
| Gene Symbol: LOC4336354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781619.1 | XP_015637105.1 | 2 - 3 | 324 | 83 | |
| GenBank mRNA | AK062117.1 | - | 347 | 83 | ||
| AK065962.1 | - | 2 - 3 | 193 | 83 | ||
More Information
Set Membership: |
|

LOC4336354
Genomic Map