Assay Details
| Gene Symbol: LOC4336384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778572.2 | XP_015634058.1 | 13 - 14 | 1966 | 71 | |
| XM_015778573.2 | XP_015634059.1 | 14 - 15 | 1814 | 71 | ||
| GenBank mRNA | AK103203.1 | - | 13 - 14 | 1963 | 71 | |
More Information
Set Membership: |
|

LOC4336384
Genomic Map