Assay Details
| Gene Symbol: LOC4336384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778572.2 | XP_015634058.1 | 5 - 6 | 996 | 146 | |
| XM_015778573.2 | XP_015634059.1 | 6 - 7 | 844 | 146 | ||
| GenBank mRNA | AK103203.1 | - | 5 - 6 | 993 | 146 | |
| Gene Symbol: LOC4336150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288722.1 | - | 1765 | 146 | ||
More Information
Set Membership: |
|

LOC4336384
Genomic Map