Assay Details
| Gene Symbol: LOC4336384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778572.2 | XP_015634058.1 | 6 - 7 | 1102 | 130 | |
| XM_015778573.2 | XP_015634059.1 | 7 - 8 | 950 | 130 | ||
| GenBank mRNA | AK103203.1 | - | 6 - 7 | 1099 | 130 | |
| Gene Symbol: LOC4336150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288722.1 | - | 1659 | 130 | ||
More Information
Set Membership: |
|

LOC4336384
Genomic Map