Assay Details
| Gene Symbol: LOC4336384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778572.2 | XP_015634058.1 | 7 - 8 | 1162 | 113 | |
| XM_015778573.2 | XP_015634059.1 | 8 - 9 | 1010 | 113 | ||
| GenBank mRNA | AK103203.1 | - | 7 - 8 | 1159 | 113 | |
| Gene Symbol: LOC4336150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288722.1 | - | 1599 | 113 | ||
More Information
Set Membership: |
|

LOC4336384
Genomic Map