Assay Details
| Gene Symbol: LOC4336429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780886.2 | XP_015636372.1 | 1 | 665 | 89 | |
| GenBank mRNA | AK105173.1 | - | 1 | 645 | 89 | |
More Information
Set Membership: |
|

LOC4336429
Genomic Map