Assay Details
| Gene Symbol: LOC4336552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781520.2 | XP_015637006.1 | 2 - 3 | 706 | 72 | |
| GenBank mRNA | AK064289.1 | - | 2 - 3 | 481 | 72 | |
More Information
Set Membership: |
|

LOC4336552
Genomic Map