Assay Details
| Gene Symbol: LOC4336554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781514.2 | XP_015637000.1 | 3 - 4 | 984 | 91 | |
| GenBank mRNA | AB189166.1 | - | 3 - 4 | 685 | 91 | |
| AK063467.1 | - | 3 - 4 | 776 | 91 | ||
| AK065301.1 | - | 2 - 3 | 2430 | 91 | ||
| AK289207.1 | - | 1241 | 91 | |||
More Information
Set Membership: |
|

LOC4336554
Genomic Map