Assay Details
| Gene Symbol: LOC4336560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777918.2 | XP_015633404.1 | 14 - 15 | 1135 | 80 | |
| XM_015777919.2 | XP_015633405.1 | 14 - 15 | 1117 | 80 | ||
| XM_015777920.2 | XP_015633406.1 | 15 - 16 | 1168 | 80 | ||
| XM_015777921.2 | XP_015633407.1 | 11 - 12 | 930 | 80 | ||
| XM_015777923.2 | XP_015633409.1 | 12 - 13 | 930 | 80 | ||
| GenBank mRNA | AK069661.1 | - | 14 - 15 | 1084 | 80 | |
More Information
Set Membership: |
|

LOC4336560
Genomic Map