Assay Details
| Gene Symbol: LOC4336560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777918.2 | XP_015633404.1 | 15 - 16 | 1204 | 93 | |
| XM_015777919.2 | XP_015633405.1 | 15 - 16 | 1186 | 93 | ||
| XM_015777920.2 | XP_015633406.1 | 16 - 17 | 1237 | 93 | ||
| XM_015777921.2 | XP_015633407.1 | 12 - 13 | 999 | 93 | ||
| XM_015777923.2 | XP_015633409.1 | 13 - 14 | 999 | 93 | ||
| GenBank mRNA | AK069661.1 | - | 15 - 16 | 1153 | 93 | |
More Information
Set Membership: |
|

LOC4336560
Genomic Map