Assay Details
| Gene Symbol: LOC4336560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777918.2 | XP_015633404.1 | 18 - 19 | 1452 | 154 | |
| XM_015777919.2 | XP_015633405.1 | 18 - 19 | 1434 | 154 | ||
| XM_015777920.2 | XP_015633406.1 | 19 - 20 | 1485 | 154 | ||
| XM_015777921.2 | XP_015633407.1 | 15 - 16 | 1247 | 154 | ||
| XM_015777923.2 | XP_015633409.1 | 16 - 17 | 1247 | 154 | ||
| GenBank mRNA | AK069661.1 | - | 18 - 19 | 1401 | 154 | |
More Information
Set Membership: |
|

LOC4336560
Genomic Map