Assay Details
| Gene Symbol: LOC4336560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777918.2 | XP_015633404.1 | 3 - 4 | 291 | 86 | |
| XM_015777919.2 | XP_015633405.1 | 3 - 4 | 291 | 86 | ||
| XM_015777920.2 | XP_015633406.1 | 4 - 5 | 324 | 86 | ||
| GenBank mRNA | AK069661.1 | - | 3 - 4 | 258 | 86 | |
More Information
Set Membership: |
|

LOC4336560
Genomic Map