Assay Details
| Gene Symbol: LOC4336952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781079.1 | XP_015636565.1 | 8 - 9 | 1907 | 67 | |
| XM_015781080.2 | XP_015636566.1 | 8 - 9 | 1968 | 67 | ||
| GenBank mRNA | AK120898.1 | - | 8 - 9 | 1909 | 67 | |
More Information
Set Membership: |
|

LOC4336952
Genomic Map