Assay Details
| Gene Symbol: LOC4336987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780781.2 | XP_015636267.1 | 2 - 3 | 756 | 134 | |
| GenBank mRNA | AK069739.1 | - | 2 - 3 | 758 | 134 | |
More Information
Set Membership: |
|

LOC4336987
Genomic Map