Assay Details
| Gene Symbol: LOC4337011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779480.2 | XP_015634966.1 | 1 - 2 | 304 | 91 | |
| GenBank mRNA | AB097461.1 | - | 1 - 2 | 34 | 91 | |
| AK060820.1 | - | 2 - 3 | 245 | 91 | ||
| AK065875.1 | - | 1 - 2 | 279 | 91 | ||
| AK102657.1 | - | 1 - 2 | 282 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015779480.2 transcripts.
Set Membership: |
|

LOC4337011
Genomic Map