Assay Details
| Gene Symbol: LOC4337011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779480.2 | XP_015634966.1 | 6 - 7 | 883 | 123 | |
| GenBank mRNA | AB097461.1 | - | 6 - 7 | 613 | 123 | |
| AK065875.1 | - | 6 - 7 | 858 | 123 | ||
| AK102657.1 | - | 6 - 7 | 861 | 123 | ||
More Information
Set Membership: |
|

LOC4337011
Genomic Map