Assay Details
| Gene Symbol: LOC4337011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779480.2 | XP_015634966.1 | 8 - 9 | 1106 | 66 | |
| GenBank mRNA | AB097461.1 | - | 8 - 9 | 836 | 66 | |
| AK060820.1 | - | 9 - 10 | 1048 | 66 | ||
| AK065875.1 | - | 8 - 9 | 1081 | 66 | ||
| AK102657.1 | - | 8 - 9 | 1084 | 66 | ||
More Information
Set Membership: |
|

LOC4337011
Genomic Map