Assay Details
| Gene Symbol: LOC4337027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779966.2 | XP_015635452.1 | 27 - 28 | 4701 | 64 | |
| XM_015779967.2 | XP_015635453.1 | 27 - 28 | 4688 | 64 | ||
| GenBank mRNA | AK240686.1 | - | 2 - 3 | 252 | 64 | |
More Information
Set Membership: |
|

LOC4337027
Genomic Map