Assay Details
| Gene Symbol: LOC4337027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779966.2 | XP_015635452.1 | 9 - 10 | 1217 | 82 | |
| XM_015779967.2 | XP_015635453.1 | 9 - 10 | 1204 | 82 | ||
More Information
Set Membership: |
|

LOC4337027
Genomic Map