Assay Details
| Gene Symbol: LOC4337190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781121.2 | XP_015636607.1 | 1 - 2 | 336 | 63 | |
| GenBank mRNA | DQ272692.1 | - | 1 - 2 | 251 | 63 | |
More Information
Set Membership: |
|

LOC4337190
Genomic Map