Assay Details
| Gene Symbol: LOC4337267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781078.2 | XP_015636564.1 | 5 - 6 | 720 | 70 | |
| GenBank mRNA | AK067399.1 | - | 5 - 6 | 717 | 70 | |
More Information
Set Membership: |
|

LOC4337267
Genomic Map