Assay Details
| Gene Symbol: LOC4337338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777975.2 | XP_015633461.1 | 3 - 4 | 542 | 124 | |
| GenBank mRNA | AK288046.1 | - | 3 - 4 | 402 | 124 | |
| AY224471.1 | - | 3 - 4 | 366 | 124 | ||
More Information
Set Membership: |
|

LOC4337338
Genomic Map