Assay Details
| Gene Symbol: LOC4337720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782361.2 | XP_015637847.1 | 5 - 6 | 529 | 143 | |
| GenBank mRNA | AB119279.1 | - | 5 - 6 | 471 | 143 | |
| AK068407.1 | - | 5 - 6 | 506 | 143 | ||
| AK099152.1 | - | 5 - 6 | 506 | 143 | ||
More Information
Set Membership: |
|

LOC4337720
Genomic Map