Assay Details
| Gene Symbol: LOC4338045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783920.2 | XP_015639406.1 | 15 - 16 | 1587 | 140 | |
| GenBank mRNA | AY858584.1 | - | 15 - 16 | 1575 | 140 | |
More Information
Set Membership: |
|

LOC4338045
Genomic Map