Assay Details
| Gene Symbol: LOC4338045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783920.2 | XP_015639406.1 | 7 - 8 | 1026 | 71 | |
| XM_015783921.2 | XP_015639407.1 | 7 - 8 | 1178 | 71 | ||
| GenBank mRNA | AY858584.1 | - | 7 - 8 | 1014 | 71 | |
More Information
Set Membership: |
|

LOC4338045
Genomic Map