Assay Details
| Gene Symbol: LOC4338045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783920.2 | XP_015639406.1 | 9 - 10 | 1256 | 75 | |
| XM_015783921.2 | XP_015639407.1 | 9 - 10 | 1408 | 75 | ||
| GenBank mRNA | AY858584.1 | - | 9 - 10 | 1244 | 75 | |
More Information
Set Membership: |
|

LOC4338045
Genomic Map