Assay Details
| Gene Symbol: LOC4338045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783920.2 | XP_015639406.1 | 13 - 14 | 1476 | 80 | |
| GenBank mRNA | AY858584.1 | - | 13 - 14 | 1464 | 80 | |
More Information
Set Membership: |
|

LOC4338045
Genomic Map