Assay Details
| Gene Symbol: LOC4338174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784081.1 | XP_015639567.1 | 38 - 39 | 4855 | 118 | |
| GenBank mRNA | AB982929.1 | - | 38 - 39 | 4738 | 118 | |
| AK120473.1 | - | 15 - 16 | 1625 | 118 | ||
More Information
Set Membership: |
|

LOC4338174
Genomic Map