Assay Details
| Gene Symbol: LOC4338174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784081.1 | XP_015639567.1 | 39 - 40 | 4999 | 96 | |
| GenBank mRNA | AB982929.1 | - | 39 - 40 | 4882 | 96 | |
| AK120473.1 | - | 16 - 17 | 1769 | 96 | ||
More Information
Set Membership: |
|

LOC4338174
Genomic Map