Assay Details
| Gene Symbol: LOC4338174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784081.1 | XP_015639567.1 | 40 - 41 | 5122 | 73 | |
| GenBank mRNA | AB982929.1 | - | 40 - 41 | 5005 | 73 | |
| AK120473.1 | - | 17 - 18 | 1892 | 73 | ||
More Information
Set Membership: |
|

LOC4338174
Genomic Map