Assay Details
| Gene Symbol: LOC4338174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784081.1 | XP_015639567.1 | 41 - 42 | 5281 | 86 | |
| GenBank mRNA | AB982929.1 | - | 41 - 42 | 5164 | 86 | |
| AK120473.1 | - | 18 - 19 | 2051 | 86 | ||
More Information
Set Membership: |
|

LOC4338174
Genomic Map