Assay Details
| Gene Symbol: LOC4338174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784081.1 | XP_015639567.1 | 49 - 50 | 6398 | 145 | |
| GenBank mRNA | AB982929.1 | - | 49 - 50 | 6281 | 145 | |
| AK120473.1 | - | 26 - 27 | 3168 | 145 | ||
More Information
Set Membership: |
|

LOC4338174
Genomic Map