Assay Details
| Gene Symbol: LOC4338174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784081.1 | XP_015639567.1 | 27 - 28 | 3645 | 72 | |
| GenBank mRNA | AB982929.1 | - | 27 - 28 | 3528 | 72 | |
| AK120473.1 | - | 4 - 5 | 415 | 72 | ||
More Information
Set Membership: |
|

LOC4338174
Genomic Map