Assay Details
| Gene Symbol: LOC4338201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782243.2 | XP_015637729.1 | 3 - 4 | 593 | 72 | |
| GenBank mRNA | AK064349.1 | - | 3 - 4 | 586 | 72 | |
More Information
Set Membership: |
|

LOC4338201
Genomic Map