Assay Details
| Gene Symbol: LOC4338448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783697.2 | XP_015639183.1 | 2 - 3 | 502 | 87 | |
| XM_015783698.2 | XP_015639184.1 | 2 - 3 | 502 | 87 | ||
| GenBank mRNA | D38232.1 | - | 1 - 2 | 145 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783698.2 transcripts.
Set Membership: |
|

LOC4338448
Genomic Map